  • Search Results
  • Transient domoic acid excitotoxicity increases BDNF expression and activates both MEK- and PKA-dependent neurogenesis in organotypic hippocampal slices.
Transient domoic acid excitotoxicity increases BDNF expression and activates both MEK- and PKA-dependent neurogenesis in organotypic hippocampal slices.
BMC neuroscience (2013-07-20)
Anabel Pérez-Gómez, R Andrew Tasker
ABSTRACT
We have previously reported evidence of cell proliferation and increased neurogenesis in rat organotypic hippocampal slice cultures (OHSC) after a transient excitotoxic injury to the hippocampal CA1 area induced by low concentrations of the AMPA/kainate agonist domoic acid (DOM). An increased baseline rate of neurogenesis may contribute to recovery from DOM-induced mild injury but the intracellular mechanism(s) responsible for neuronal proliferation remain unclear. The current study investigated the key intracellular pathways responsible for DOM-induced neurogenesis in OHSC including the effects of transient excitotoxicity on the expression of brain-derived neurotrophic factor (BDNF), a well-known regulator of progenitor cell mitosis. Application of a low concentration of DOM (2 μM) for 24 h followed by recovery induced a significant and long lasting increase in BDNF protein levels expressed by both neurons and microglial cells. Furthermore, the mild DOM toxicity stimulated both PKA and MEK-dependent intracellular signaling cascades and induced a significant increase in BDNF- transcription factor CREB activation and BDNF-receptor TrkB expression. Coexposure to specific inhibitors of PKA and MEK phosphorylation resulted in a significant decrease in the neurogenic marker doublecortin. Our results suggest that transient excitotoxic insult induced by DOM produces BDNF and CREB overexpression via MEK and PKA pathways and that both pathways mediate, at least in part, the increased neural proliferation resulting from mild excitotoxicity.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Brain Derived Neurotrophic Factor Antibody, serum, Chemicon®