  • Search Results
  • Vps34 regulates Rab7 and late endocytic trafficking through recruitment of the GTPase-activating protein Armus.
Vps34 regulates Rab7 and late endocytic trafficking through recruitment of the GTPase-activating protein Armus.
Journal of cell science (2016-11-02)
Nadia Jaber, Noor Mohd-Naim, Ziqing Wang, Jennifer L DeLeon, Seong Kim, Hua Zhong, Namratha Sheshadri, Zhixun Dou, Aimee L Edinger, Guangwei Du, Vania M M Braga, Wei-Xing Zong
ABSTRACT
The class III phosphoinositide 3-kinase (PI3K) Vps34 (also known as PIK3C3 in mammals) produces phosphatidylinositol 3-phosphate [PI(3)P] on both early and late endosome membranes to control membrane dynamics. We used Vps34-deficient cells to delineate whether Vps34 has additional roles in endocytic trafficking. In Vps34
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid