  • Search Results
  • Targeting metastasis-initiating cells through the fatty acid receptor CD36.
Targeting metastasis-initiating cells through the fatty acid receptor CD36.
Nature (2016-12-16)
Gloria Pascual, Alexandra Avgustinova, Stefania Mejetta, Mercè Martín, Andrés Castellanos, Camille Stephan-Otto Attolini, Antoni Berenguer, Neus Prats, Agustí Toll, Juan Antonio Hueto, Coro Bescós, Luciano Di Croce, Salvador Aznar Benitah
ABSTRACT
The fact that the identity of the cells that initiate metastasis in most human cancers is unknown hampers the development of antimetastatic therapies. Here we describe a subpopulation of CD44
Sigma-Aldrich
Complete Whole Transcriptome Amplification Kit, DNA polymerase included, Complete Kit with optimized enzyme to amplify total RNA in <4 hours, no 3′ bias
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%