Combined treatment with a β
Neurourology and urodynamics (2016-07-02)
Tetsuya Imamura, Teruyuki Ogawa, Tomonori Minagawa, Takashi Nagai, Toshiro Suzuki, Tetsuichi Saito, Hitoshi Yokoyama, Masaki Nakazawa, Osamu Ishizuka
ABSTRACT
This study determined if combined treatment with the muscarinic receptor (MR) antagonist solifenacin and the β Thirty-two female 10-week-old SHRs were fed an 8% NaCl-supplemented diet for 4 weeks. Cystometric measurements of the unanesthetized, unrestricted rats were performed at room temperature (RT, 27 ± 2°C) for 20 min. The rats were then intravenously administered vehicle, 0.1 mg/kg solifenacin alone, 0.1 mg/kg mirabegron alone, or the combination of 0.1 mg/kg mirabegron and 0.1 mg/kg solifenacin (n = 8 each group). Five minutes later, the treated rats were exposed to low temperature (LT, 4 ± 2°C) for 40 min. Finally, the rats were returned to RT. After the cystometric investigations, the β Just after transfer from RT to LT, vehicle-, solifenacin-, and mirabegron-treated SHRs exhibited detrusor overactivity that significantly decreased voiding interval and bladder capacity. However, treatment with the combination of solifenacin and mirabegron partially inhibited the cold stress-induced detrusor overactivity patterns. The decreases of voiding interval and bladder capacity in the combination-treated rats were significantly inhibited compared to other groups. Within the urinary bladders, there were no differences between expression levels of M This study suggested that the combination of solifenacin and mirabegron act synergistically to inhibit the cold stress-induced detrusor overactivity in SHRs. Neurourol. Urodynam. 36:1026-1033, 2017. © 2016 The Authors. Neurourology and Urodynamics Published by Wiley Periodicals, Inc.
Sigma-Aldrich
Solketal methacrylate, 50 wt. % in dichloromethane, contains ~280 ppm 4-tert-butylcatechol as inhibitor