  • Search Results
  • Atmospheric protein chemistry influenced by anthropogenic air pollutants: nitration and oligomerization upon exposure to ozone and nitrogen dioxide.
Atmospheric protein chemistry influenced by anthropogenic air pollutants: nitration and oligomerization upon exposure to ozone and nitrogen dioxide.
Faraday discussions (2017-06-03)
Fobang Liu, Pascale S J Lakey, Thomas Berkemeier, Haijie Tong, Anna Theresa Kunert, Hannah Meusel, Yafang Cheng, Hang Su, Janine Fröhlich-Nowoisky, Senchao Lai, Michael G Weller, Manabu Shiraiwa, Ulrich Pöschl, Christopher J Kampf
ABSTRACT
The allergenic potential of airborne proteins may be enhanced via post-translational modification induced by air pollutants like ozone (O
Product Description
Supelco
Protein Standard Mix 15 - 600 kDa, for size exclusion chromatography
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)