Merck
Establishment of mouse expanded potential stem cells.
Nature (2017-10-12)
Jian Yang, David J Ryan, Wei Wang, Jason Cheuk-Ho Tsang, Guocheng Lan, Hideki Masaki, Xuefei Gao, Liliana Antunes, Yong Yu, Zhexin Zhu, Juexuan Wang, Aleksandra A Kolodziejczyk, Lia S Campos, Cui Wang, Fengtang Yang, Zhen Zhong, Beiyuan Fu, Melanie A Eckersley-Maslin, Michael Woods, Yosuke Tanaka, Xi Chen, Adam C Wilkinson, James Bussell, Jacqui White, Ramiro Ramirez-Solis, Wolf Reik, Berthold Göttgens, Sarah A Teichmann, Patrick P L Tam, Hiromitsu Nakauchi, Xiangang Zou, Liming Lu, Pentao Liu
ABSTRACT
Mouse embryonic stem cells derived from the epiblast contribute to the somatic lineages and the germline but are excluded from the extra-embryonic tissues that are derived from the trophectoderm and the primitive endoderm upon reintroduction to the blastocyst. Here we report that cultures of expanded potential stem cells can be established from individual eight-cell blastomeres, and by direct conversion of mouse embryonic stem cells and induced pluripotent stem cells. Remarkably, a single expanded potential stem cell can contribute both to the embryo proper and to the trophectoderm lineages in a chimaera assay. Bona fide trophoblast stem cell lines and extra-embryonic endoderm stem cells can be directly derived from expanded potential stem cells in vitro. Molecular analyses of the epigenome and single-cell transcriptome reveal enrichment for blastomere-specific signature and a dynamic DNA methylome in expanded potential stem cells. The generation of mouse expanded potential stem cells highlights the feasibility of establishing expanded potential stem cells for other mammalian species.
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
XAV939, ≥98% (HPLC)
Sigma-Aldrich
Galantide, ≥90% (HPLC)
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Tyrode′s Solution, Acidic, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
M2 medium, With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
IWR-1, ≥98% (HPLC)