Merck
  • Allosteric modulation of ligand binding to [3H](+)pentazocine-defined sigma recognition sites by phenytoin.
Allosteric modulation of ligand binding to [3H](+)pentazocine-defined sigma recognition sites by phenytoin.
Life sciences (1993-01-01)
D L DeHaven-Hudkins, F Y Ford-Rice, J T Allen, R L Hudkins
ABSTRACT
The allosteric modulation of sigma recognition sites by phenytoin (diphenylhydantoin) has been demonstrated by the ability of phenytoin to stimulate binding of various [3H] sigma ligands, as well as to slow dissociation from sigma sites and to shift sigma sites from a low- to a high-affinity state. Phenytoin stimulated the binding of the sigma 1- selective ligand [3H](+)pentazocine in a dose-dependent manner. Stimulation of binding at a final concentration of 250 microM phenytoin was associated with a decrease in the KD. The affinities of the sigma reference compounds caramiphen, dextromethorphan, dextrophan, (+)3-PPP and (+)SKF-10,047 were three- to eight-fold higher, while the affinities of benzetimide, BMY-14802, carbetapentane, DTG and haloperidol were unchanged in the presence of 250 microM phenytoin. The relative sensitivity of sigma compounds to allosteric modulation by phenytoin is not a property of all sigma ligands, and may provide an in vitro basis for distinguishing actions of sigma compounds and predicting sigma effects in vivo.
Sigma-Aldrich
Caramiphen hydrochloride, ≥98% (HPLC)