Merck
Leptin Induces Sca-1
Arteriosclerosis, thrombosis, and vascular biology (2017-09-25)
Yao Xie, Claire M F Potter, Alexandra Le Bras, Witold N Nowak, Wenduo Gu, Shirin Issa Bhaloo, Zhongyi Zhang, Yanhua Hu, Li Zhang, Qingbo Xu
ABSTRACT
Leptin is an adipokine initially thought to be a metabolic factor. Recent publications have shown its roles in inflammation and vascular disease, to which Sca-1 Sca-1 Upregulation of leptin levels in both the vessel wall and the circulation after vessel injury promoted the migration of Sca-1
Sigma-Aldrich
Solketal methacrylate, 50 wt. % in dichloromethane, contains ~280 ppm 4-tert-butylcatechol as inhibitor
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture