  • Search Results
  • Polythiophene/graphene oxide nanostructured electrodeposited coating for on-line electrochemically controlled in-tube solid-phase microextraction.
Polythiophene/graphene oxide nanostructured electrodeposited coating for on-line electrochemically controlled in-tube solid-phase microextraction.
Journal of chromatography. A (2016-11-12)
Maryam Shamsayei, Yadollah Yamini, Hamid Asiabi
ABSTRACT
In this work, a novel polythiophene/graphene oxide (PTh/GO) nanostructured coating was introduced for on-line electrochemically-controlled in-tube solid phase microextraction of amitriptyline (AMI) and doxepin (DOX) as antidepressant drugs. The PTh/GO coating was prepared on the inner surface of a stainless steel tube by a facile in-situ electro-deposition method and it was used as a working electrode for electrochemically control in-tube solid phase microextraction. In the PTh/GO coating, GO acts as an anion dopant and sorbent. The PTh/GO coating, compared to PTh and GO coatings, exhibited enhanced long lifetime, good mechanical stability and a large specific surface area. Regarding the in-tube SPME, some important factors such as the extraction and desorption voltage, extraction and desorption times and flow rates of the sample solution and eluent, which could affect the extraction and separation efficiency of the analytes, were optimized. Total analysis time of this method including the online extraction and desorption time was about 21min for each sample. AMI and DOX were extracted, separated and determined with limits of detection as small as 0.3μgL
Product Description
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm (CAR/PDMS), needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm (CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm (PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 23 ga, for use with manual holder
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm (CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with autosampler, needle size 24 ga
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), for use with manual holder, needle size 24 ga
Supelco
SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 75 μm (Coating thickness includes 65 μm coating + 10 μm OC (overcoating)), needle size 23 ga, pkg of 3 ea, fiber L 1 cm