Documents
Z155233
Sigma-Aldrich
Solid cover for type 304 SS pan
Fits pans 20.75 in. x 12.75 in.
material
stainless steel cover
manufacturer/tradename
Polarware 2000-2
L × W
20.75 in. × 12.75 in.
capacity
8.5, 14.5 and 24 qt