Documents
Z160407
Fluke® Type-K immersion probe
temperature -40-1800 °F
manufacturer/tradename
Fluke
parameter
-40-1800 °F temperature
General description
For liquids and gels. Protected by Inconel sheath, can be used in most general temperature measurement applications.
Legal Information
Fluke is a registered trademark of John Fluke Manufacturing Co., Inc.