Documents
Z162450
Economy crucible and flask tongs
stainless steel, L 9 in. (229 mm)
NACRES:
NB.43
material
stainless steel
L
9 in. (229 mm)
General description
All-purpose laboratory and industrial tong for holding, manipulating and pouring, as well as for picking up hot objects. With serrated tips.