Documents
Z181692
Supelco
Bottom-drip adapter for large columns
Luer tip, 1 mm bore, Nylon
NACRES:
SB.52
material
Nylon
manufacturer/tradename
Ace Glass 583721
application(s)
LPLC: suitable
Looking for similar products? Visit Product Comparison Guide
General description
Shallow taper of upper end minimizes mixing below the PE packing support disc (included). All adapters have an Ace Thred #50 and include one FETFE® O-ring.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.