Documents
Z341711
3M 6000/7000 series filters
P3 Particulate filter, pkg of 2 ea
NACRES:
NB.35
description
Protects against particulates & non-volatile mists up to 50 × OEL, EN143
packaging
pkg of 2 ea
manufacturer/tradename
3M 2135
availability
not available in USA and Canada
Looking for similar products? Visit Product Comparison Guide
General description
Gas and vapour filters
Two filters required per respirator.
Features and Benefits
These filters utilise 3M′s advanced filter technology which minimises breathing resistance for greater comfort to the user. Select gas and vapour filters, particulate filters according to protection required. All filters fit both 6000 and 7000 series respirators by bayonet connection. Two filters required per respirator.
Warning
All these filters should only be used for protection against hazards with good warning properties, e.g. smell or taste, and providing the concentrations of hazardous substances are not immediately dangerous to life or health (IDLH). *Where concentrations are given in terms of the OEL or ppm, the filter should only be used for protection against concentrations below the lower value.
Legal Information
3M is a trademark of 3M Company
Certificate of Analysis
Certificate of Origin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.