Documents
Z619256
Heidolph Rotary Evaporator, Laborota 4010
vertical condenser G3B, plastic-coated glass
AC/DC input
230 V
material
plastic-coated glass
feature
CE compliant
parameter
20-180 °C temperature
speed
20-270 rpm
Looking for similar products? Visit Product Comparison Guide
General description
Safety features
  • For your protection against splashing water and steam, the operating panel is located in front of the evaporator for safe and easy access.
  • Drip shield over display protects user and control panel from spills
  • Individual power switches for control panel and heating bath offers additional safety
  • Heating bath features non-slip safety handles for safe and easy transport of bath
  • Heating bath is constructed of an insulated double wall for you protection against burns and scalding
  • Adjustable sliding safety guard hood always aligns with the bath and protects you in case of implosion
Convenience features
  • Long life graphite filled PTFE vacuum seal
  • Non-sticking vapor tube removes easily eliminating broken glass
  • Unique large bore vapor tube offers fast distribution rates
  • One piece vapor tube without additional sealing
  • Flask ejector presses off sticking evaporation flasks
  • Adjust and fix the height of your evaporating flask easily
  • For your convenience, the angle of the evaporation flask can be adjusted from 10 ° to 85 ° quickly
  • Superior results due to high torque (140 Ncm) drive
  • Choice of manual or motor lift
Certificate of Analysis
Certificate of Origin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.