EHU090701
Sigma-Aldrich
MISSION® esiRNA
targeting human SPATA33 (1)
NACRES:
NA.51
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
GGCAAGAGTGACTGGCTCAGAGAGCGAGCTTTGTGGTATTGATAAATGAATAAATTAGTATGTTCATGTTGGAAGTGGCTGGCTCTGCAGAGTGATGGACAGCAGGTGGCATCTTACATTGTGATTTGTGGTGCACAGTTAAAACACTTCCCTTCGTTTCAGAAGGAGACAATGTGAATACTTGGAAATTATAGTGAATAAGATGCCTCCCAAACTTGAGGAAAAGGTCAGATTCTAAGAGCTAAACGTTAAGATGTAATGTTCCAGAACAAAGAGGCCCAGAGTCACTGAACGGGGCCCCAGGGCTCCTAGCTTGAGTGTAATCTGAGTCAGCTTATGAAAAGGCCCCAAAGAAGAAAGTGGAATGTGTGCTGGGCACATGGGACAGCCATGGCACTGA
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... SPATA33(124045), C16orf55(124045)
NONH for all modes of transport
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable