EHU101611
Sigma-Aldrich
MISSION® esiRNA
targeting human TRAV19
NACRES:
NA.51
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
TTGTATCCAGCATGGCTCAGAAGGTAACTCAAGCGCAGACTGAAATTTCTGTGGTGGAGAAGGAGGATGTGACCTTGGACTGTGTGTATGAAACCCGTGATACTACTTATTACTTATTCTGGTACAAGCAACCACCAAGTGGAGAATTGGTTTTCCTTATTCGTCGGAACTCTTTTGATGAGCAAAATGAAATAAGTGGTCGGTATTCTTGGAACTTCCAGAAATCCACCAGTTCCTTCAACTTCACCATCACAGCCTCACAAGTCGTGGACTCA
Ensembl | human accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... AE000660.1(6256)
NONH for all modes of transport
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable