T1914
Sigma-Aldrich
Silicone rubber laboratory tubing
I.D. × O.D. 3/32  in. × 5/32  in.
NACRES:
NB.35
I.D. × O.D.
3/32  in. × 5/32  in.
wall size
1/32  in.
Looking for similar products? Visit Product Comparison Guide
General description
Translucent and flexible. Temp. range: -60 to 460°F (-51 to 238°C). Do not use with concentrated acids or dilute sodium hydroxide. Suitable for high-temperature applications.