Documents
Z660264
Ohaus® Adventurer® Pro balance
Ohaus AV4101, AC/DC input 230 V AC
Synonym(s):
analytical scales, weighing balances, laboratory balances
feature
CE compliant
CSA approved
manufacturer/tradename
Ohaus AV4101
availability
available only in EU
weighing capacity measuring range
4,100 g, resolution: 0.1 g
platform L × W
149 mm × 162 mm
Looking for similar products? Visit Product Comparison Guide
General description
  • Brilliant backlit high resolution dot matrix display
  • RS232 port for easy communication with computer or printer
  • GLP compliant with user selectable outputs
  • AC adapter (supplied) or 4 "AA" batteries (not supplied)
  • Sealed front panel and moulded spill ring to protect the balance during use
  • Includes an integral shipping lock to protect the weighing mechanism during transportation
  • Lockswitch to allow balance to be locked into a specific configuration, reducing operator error
  • Industry leading warranty and support
Legal Information
Adventurer is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Certificate of Analysis
Certificate of Origin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.