Documents
Z660345
Ohaus® Explorer® Pro balance
EP4102C, AC/DC input 230 V AC
feature
CE compliant
CSA approved
manufacturer/tradename
EP4102C
availability
available only in EU
weighing capacity measuring range
4,100 g, resolution: 10 mg
platform L × W
172 mm × 172 mm
Looking for similar products? Visit Product Comparison Guide
General description
The Explorer Pro Series of standard level balances was designed for simplicity and includes the industry′s easiest user interface for easy setup and operation; no training is required.
  • Simple 2 key menu operation and navigation
  • Text prompts for fast setup
  • 7 Application modes plus below balance weighing
  • Brilliant backlit high resolution dot matrix display
  • RS232 port for easy communication with computer or printer
  • GLP compliant with user selectable outputs
  • 5 Language operational display text
  • Protective in-use cover and security bracket included
  • Industry leading warranty and support
Legal Information
Explorer is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Certificate of Analysis
Certificate of Origin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.