58764
Plug
for 1/16 in. tubing
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
SSI
fitting
1/4-28 plug
General description
The stainless steel plug is used for connecting most columns to a 1/16 in. tubing system.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis