PS355

Propachlor

analytical standard

Synonym(s):
2-Chloro-N-isopropylacetanilide
Empirical Formula (Hill Notation):
C11H14ClNO
CAS Number:
Molecular Weight:
211.69
Beilstein/REAXYS Number:
2103903
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

form

neat

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-355

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Agriculture
Environmental

format

neat

SMILES string

CC(C)N(C(=O)CCl)c1ccccc1

InChI

1S/C11H14ClNO/c1-9(2)13(11(14)8-12)10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3

InChI key

MFOUDYKPLGXPGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN2811 - class 6.1 - PG 3 - MP - EHS - Toxic solids, organic, n.o.s., HI: all

WGK Germany

3

Flash Point(F)

212 °F

Flash Point(C)

100 °C

Certificate of Analysis
Certificate of Origin
Sheng Tai et al.
Molecular cancer therapeutics, 11(6), 1320-1331 (2012-04-12)
PI3K/AKT/mTOR pathway plays a key role in the tumorigenesis of many human cancers including prostate cancer. However, inhibitors of this pathway, such as Rad001, have not shown therapeutic efficacy as a single agent. Through a high-throughput screen of 5,000 widely...
A R Loch et al.
Environmental science & technology, 36(19), 4065-4073 (2002-10-17)
Reactions of bisulfide and polysulfides with alachlor, propachlor, and metolachlor were examined in aqueous solution to investigate the role reduced sulfur species could play in effecting abiotic transformations of chloroacetanilide herbicides. Experiments at 25 degrees C demonstrated that reactions were...
M B Genter et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 47(6), 1051-1057 (2009-05-09)
Alachlor and butachlor are chloracetanilide herbicides that induce olfactory tumors in rats, whereas propachlor does not. The mechanism by which alachlor induces tumors is distinct from many other nasal carcinogens, in that alachlor induces a gradual de-differentiation of the olfactory...
K L Davison et al.
Xenobiotica; the fate of foreign compounds in biological systems, 24(10), 1003-1012 (1994-10-01)
1. 14C-labelled propachlor, alachlor, butachlor, metolachlor, methoxypropachlor and some of their mercapturic acid pathway metabolites (MAP) were given to rat either by gavage or by perfusion into a renal artery. MAP metabolites were isolated from bile and urine. 2. Rat...
Ioannis K Konstantinou et al.
Water research, 36(11), 2733-2742 (2002-07-31)
The light-induced degradation of propachlor (2-chloro-N-isopropylacetanilide) under simulated solar irradiation has been investigated in aqueous solutions containing TiO2 suspensions as photocatalysts. The study focuses on the identification of possible intermediate products and the determination of inorganic ions formed during the...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.