Product Name
(2-Hydroxypropyl)-α-cyclodextrin average Mw ~1,180
Product Number
390690
Product Brand
ALDRICH
CAS Number
128446-33-3
Molecular Weight
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
OPTICAL ROTATION119.5 - 130.5 Degrees
CONCENTRATIONC=1, H2O
INFRARED SPECTRUMCONFORMS TO STRUCTURE
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE