Product Name
Scandium(III) triflate 99.995% trace metals basis
Product Number
483354
Product Brand
ALDRICH
CAS Number
144026-79-9
Molecular Weight
492.16
Test
Specification
Appearance (Color)White
Appearance (Form)Conforms to Requirements
 Powder, Crystals and/or Chunks
Infrared spectrumConforms to Structure
ICP: Confirms Scandium ComponentConfirmed
Trace Rare Earth Analysis≤60.0 ppm
Boron (B)ppm
Barium (Ba)ppm
Calcium (Ca)ppm
Iron (Fe)ppm
Lutetium (Lu)ppm
Magnesium (Mg)ppm
Sodium (Na)ppm
Zinc (Zn)ppm
PurityMeets Requirements
 99.995% Based On Trace Metals Analysis