Product Name
Aluminum oxide nanopowder, <50 nm particle size (TEM)
Product Number
544833
Product Brand
ALDRICH
CAS Number
1344-28-1
Molecular Weight
101.96
Test
Specification
Appearance (Color)White
Appearance (Form)Powder
ICP: Confirms Aluminum ComponentConforms
Size≤50 nm