Product Name
Boc-Trp(For)-PAM resin extent of labeling: ~0.6 mmol/g loading
Product Number
56932
Product Brand
ALDRICH
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)White to Brown
APPEARANCE (FORM)Powder or Crystals
PARTICLE SIZE100 - 200 MESH
CARBON CONTENT--
HYDROGEN CONTENT--
NITROGEN CONTENT--
MISCELLANEOUS TESTSLOADING: ~0.6 MMOL/G