Product Name
TentaGel TBTA extent of labeling: 0.17 mmol/g loading
Product Number
696773
Product Brand
ALDRICH
CAS Number
Molecular Weight
Test
Specification
APPEARANCEWHITE TO YELLOW SOLID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSIS~1.90 MMOL/G NITROGEN
MASS BALANCE~0.17 MMOL TBTA/G
 APPROVED APRIL 21, 2008 LUKE GROCHOLL