Product Name
Spinosad PESTANAL®, analytical standard
Product Number
33706
Product Brand
SIAL
CAS Number
168316-95-8
Molecular Weight
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
MELTING POINT137 - 144 C
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS