Product Name
Deltamethrin PESTANAL®, analytical standard
Product Number
45423
Product Brand
SIAL
CAS Number
52918-63-5
Molecular Weight
505.20
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
SPECIFIC ROTATION (20/D)19.0 - 25.0 Degrees
CONCENTRATIONC=1.4 in CHLOROFORM
MELTING POINT98 - 103 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS