Product Name
4-Methyl-5-(sulfomethylamino)-2-(2-thiazolylazo)benzoic acid for spectrophotometric det. of Co(II), Cu(II), ≥90%
Product Number
87793
Product Brand
SIAL
CAS Number
82138-69-0
Molecular Weight
356.38
Test
Specification
APPEARANCE (COLOR)Light Blue to Very Dark Blue and Black
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 90.0 %
ASSAY (ELEMENTAL ANALYSIS)≥ 90.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
APPLICATION TESTfor spectrophotometric det. of Co(II), Cu(II)