Product Name
Terbuthylazine-(ethyl-d5) analytical standard
Product Number
91799
Product Brand
SIAL
CAS Number
222986-60-9
Molecular Weight
234.74
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (GC AREA %)≥ 98.0 %
IDENTIFICATION (GC-MS)CORRESPONDS TO REQUIREMENTS
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
CARBON 13 NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS