Product Name
MISSION® esiRNA targeting human RSF1
Product Number
EHU059911
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Concentration200 ng/ul
Appearancecolorless liquid
PCR Gel Electrophoresispass
PCR DNA Sequencingpass
Concentration/OD260 or Fluorescence Intensitypass