Product Name
MISSION® esiRNA targeting human AGBL5
Product Number
EHU071511
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration200 ng/µL
PCR Product - DNA SequencingPass
Concentration - OD260 or Fluorescence IntensityPass