Product Name
MISSION® esiRNA targeting human LRWD1
Product Number
EHU130861
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
AppearanceColorless liquid
Concentration200 ng/µl
PCR Product - DNA SequencingPass
Concentration - OD260 or Fluorescence IntensityPass