Product Name
MISSION® esiRNA targeting human AP2S1
Product Number
EHU135191
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration200 ng/ul
Purified RNaseIII-Gel Electrophoresispass
PCR Product-DNA Sequencingpass
Concentration-OD260 or Fluorescence Intensitypass
Purified RNaseIII-Gel Electrophoresispass