Product Name
MISSION® esiRNA targeting human UGT2B11
Product Number
ehu105501
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration200 ng/µL
PCR Product - DNA SequencingPass
Concentration - OD260 or Fluorescence IntensityPass