Product Name
MISSION® esiRNA targeting human RPS7
Product Number
ehu110521
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration200 ng/ul
PCR Product-Gel Electrophoresispass
PCR Product-DNA Sequencingpass
Concentration-OD260 or Fluorescence Intensitypass
Purified RNaseIII-Gel Electrophoresispass