Product Name
MISSION® esiRNA targeting human BTBD7 (4)
Product Number
ehu117371
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration200 ng/µL
PCR Product - DNA SequencingPass
Concentration - OD260 or Fluorescence IntensityPass