  • Search Results
  • SRPX2 promotes cell proliferation and invasion via activating FAK/SRC/ERK pathway in non-small cell lung cancer.
SRPX2 promotes cell proliferation and invasion via activating FAK/SRC/ERK pathway in non-small cell lung cancer.
Acta biochimica Polonica (2020-06-19)
Xiujuan Li, Jing Liu, Hong Sun, Yong Zou, Juan Chen, Yichun Chen, Cheng Chen, Xuan Wu
ABSTRACT
Recent studies showed that sushi repeat containing protein X linked 2 (SRPX2) could participate in the development of various malignant tumors. However, its role in non-small cell lung cancer (NSCLC) was unknown. The aim of the study was to prospectively investigate the role of SRPX2 in NSCLC cell proliferation, migration and invasion and reveal the underlying mechanism. Quantitative real-time polymerase chain reaction (qRT-PCR), western blot and immunohistochemistry - IHC) were used to measure detect the mRNA and protein levels, respectively, in NSCLC tissues and cell lines. Cell Counting Kit-8 (CCK-8), colony formation, wound healing and transwell assays were utilized to assess cell proliferation, migration and invasion. In vivo subcutaneous xenograft tumor model was established to detect the tumorigenic function of SRPX2, and IHC assay was performed to measure protein expression. SRPX2 expression was upregulated in NSCLC tissues and cell lines, and positively correlated with tumor size, lymph node metastasis, distant metastasis and clinical stage. High SRPX2 expression also predicted poor prognosis. In vitro experiments indicated that overexpression of SRPX2 promoted the proliferation, migration, and invasion of SPC-A1 cells while knockdown of SRPX2 caused the opposite effects in A549 cells. Specifically, SRPX2 activated FAK/SRC/ERK pathway and its downstream effectors and promoted epithelial-mesenchymal transition (EMT). Taken together, our findings revealed a functional role of SRPX2 in NSCLC cell proliferation, migration and invasion. The underlying mechanism was, at least partially, the activation of FAK/SRC/ERK pathway. This study provides the molecular basis for targeting SRPX2 in potential clinical application for NSCLC.
Sigma-Aldrich
Anti-phospho-SRC (pTyr529) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
MISSION® esiRNA, targeting human SRPX2
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.