  • Search Results
  • [Quantitative method for simultaneous assay of four coumarins with one marker in Fraxini Cortex].
[Quantitative method for simultaneous assay of four coumarins with one marker in Fraxini Cortex].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2011-10-29)
Weihong Feng, Zhimin Wang, Qiwei Zhang, Limei Liu, Jinyu Wang, Fei Yang
ABSTRACT
To establish a new quantitative method for simultaneous determination of multi-coumarins in Fraxini Cortex by using one chemical reference substance, and validate its feasibilities. The new quality evaluation method, quantitative analysis of multi-components by singer-marker (QAMS), was established and validated with Fraxini Cortex. Four main coumarins were selected as analytes to evaluate the quality and their relative correlation factors (RCF) were determined by HPLC-DAD. Within the linear range, the values of RCF at 340 nm of aesculin to asculetin, fraxin and fraxetin were 1.771, 0.799, 1.409, respectively. And the contents of aesculin in samples of Fraxini Cortex were authentically determined by the external standard method, and the contents of the three other coumarins were calculated by their RCF. The contents of these four coumarins in all samples were also determined by the external standard method. Within a certain range, the RCF had a good reproducibility (RSD 2.5%-3.9%). Significant differences were not observed between the quantitative results of two methods. It is feasible and suitable to evaluate the quality of Fraxini Cortex and its Yinpian by QAMS.
Product Description
Supelco
Fraxin, analytical standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.