Merck
All Photos(2)

613444

Sigma-Aldrich

Deuterium oxide

99.9 atom % D, glass distilled

Synonym(s):
Heavy water, Water-d2
Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.21

Quality Level

isotopic purity

99.9 atom % D

form

liquid

technique(s)

NMR: suitable

bp

101.4 °C (lit.)

mp

3.8 °C (lit.)

SMILES string

[2H]O[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Deuterium oxide (D2O) also known as heavy water is deuterated water. It is safe and inexpensive source of deuterium (D) for the D/H exchange reactions. The equilibrium constant for the exchange of deuterium between hydrogen (H) and D2O molecules has been determined. Reports suggest that gelatin gels prepared in D2O show higher rigidity than gels prepared in water. The influence of temperature on the heat capacity of deuterium in crystalline and liquid state was studied calorimetrically and its melting point, enthalpy and entropy of fusion were determined. Its utility as a potential therapeutic agent in treating human pancreatic cancer has been studied.

Application

Deuterium oxide may be used in the following processes:
  • As an NMR solvent.
  • Preparation of β-deuterated alcohols.
  • For the adsorption of neutrons in atomic reactors.
  • Deuterium transfer agent in combination with hexamethyldisilane in the preparation of 1,2-dideuterioalkenes from alkynes.

Packaging

100 g in glass bottle

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service