Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
13CC7H8O4
Molecular Weight:
169.14
UNSPSC Code:
12352005

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+1

Related Categories

Application

- High-resolution NMR spectroscopy
- Metabolomic profiling and pathway analysis
- Stable Isotope probing in Environmental microbiology
- Acid-catalyzed reaction mechanism studies

Features and Benefits

Features
  • Versatile use in different formulations involving Vanillic acid-(methoxy-13C) ≥99 atom % 13C, ≥98% (CP).
  • Exceptional stability and compatibility with various ingredients.
  • High purity of Vanillic acid-(methoxy-13C) ≥99 atom % 13C, ≥98% (CP) ensures optimal performance.
  • Cost-effective solution for Vanillic acid-(methoxy-13C) ≥99 atom % 13C, ≥98% (CP) applications.


Benefits
  • Enhances product efficacy and reliability in formulations.
  • Improves overall product quality and consumer satisfaction.
  • Reduces time to market for products utilizing Vanillic acid-(methoxy-13C) ≥99 atom % 13C, ≥98% (CP).
  • Increases market competitiveness of products containing Vanillic acid-(methoxy-13C) ≥99 atom % 13C, ≥98% (CP).

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service