Skip to Content
Merck

BL3H9A62557C

BLD Pharmatech

α-D-Glucopyranoside, β-D-fructofuranosyl, polymer with 2-(chloromethyl)oxirane

Synonym(s):

(2R,3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol--2-(chloromethyl)oxirane (1/1)

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C12H22O11.C3H5ClO/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12;4-1-3-2-5-3/h4-11,13-20H,1-3H2;3H,1-2H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+;/m1./s1

Inchi Key

IACFXVUNKCXYJM-AKSHDPDZSA-N

Shipping Temperature

Normal

Purity

98%400000

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service