Merck
All Photos(2)

1.08484

SAFC

Urea

beads

Synonym(s):
Carbamide, Carbonyl diamide, Diaminomethanone, Carbonyldiamine, Carbamide, Carbonyldiamide
Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
MDL number:
EC Index Number:
200-315-5

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

beads
solid

technique(s)

titration: suitable

pH

9 (20 °C, 100 g/L in H2O)

mp

132-135 °C (lit.)

solubility

1000 g/L

density

1.335 g/mL at 25 °C (lit.)

application(s)

pharma/biopharma processes

storage temp.

2-30°C

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Urea is typically used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. It is useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. Urea may be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.
Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Please enter the Product Number and Lot Number.

Certificate of Quality

Please enter the Product Number and Lot Number.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service