Merck
All Photos(1)

5030

Sigma-Aldrich

Light Diagnostics Measles Antibody IFA Reagent, ~50 tests, included in kit #3187

from mouse

eCl@ss:
32161000

Quality Level

biological source

mouse

clone

monoclonal

manufacturer/tradename

Chemicon®
Light Diagnostics

technique(s)

immunofluorescence: suitable

detection method

fluorometric

shipped in

wet ice

General description

Diagnostics Kit

Application

Use undiluted. Refer to Kit Package Insert for directions.

Physical form

2 mL dropper vial containing anti-Measles in PBS with Tween 20 and Sodium Azide as a preservative.
Format: Purified

Storage and Stability

When stored at 2-8°C and protected from light, this reagent is stable up to the expiration date printed on the label. Do not freeze or expose to elevated temperatures.

Warnings / Precautions:

Sodium Azide can form potentially explosive metal azides with lead or copper plumbing. On disposal, flush with large volumes of water to prevent azide build up.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For in vitro Diagnostic Use

CE Mark

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Tyler J Wenzel et al.
Molecular and cellular neurosciences, 105, 103493-103493 (2020-04-26)
Neuroinflammation contributes to neurodegenerative disorders, including Alzheimer's disease (AD). Gut microbes are involved in regulating systemic inflammation. Short-chain fatty acids (SCFAs), which are among the many metabolites released by gut microbes, can cross the blood-brain barrier (BBB) and interact with
Jessica Hendy et al.
Nature communications, 9(1), 4064-4064 (2018-10-05)
The analysis of lipids (fats, oils and waxes) absorbed within archaeological pottery has revolutionized the study of past diets and culinary practices. However, this technique can lack taxonomic and tissue specificity and is often unable to disentangle signatures resulting from
Herlinda Clement et al.
Toxins, 11(12) (2019-12-08)
Bothropic venoms contain enzymes such as metalloproteases, serine-proteases, and phospholipases, which acting by themselves, or in synergism, are the cause of the envenomation symptoms and death. Here, two mRNA transcripts, one that codes for a metalloprotease and another for a
Isolation of measles virus from clinical specimens using B95a and Vero/hSLAM cell-lines.
Keniscope C, Juliana R, Subri H, Shangari SR, Wan Nor Azlina WA, Hamizah A, Emmi EE et al.
The Medical Journal of Malaysia null
Stefan Lambert et al.
The ISME journal, 13(2), 388-401 (2018-09-27)
Seasonality in marine microorganisms has been classically observed in phytoplankton blooms, and more recently studied at the community level in prokaryotes, but rarely investigated at the scale of individual microbial taxa. Here we test if specific marine eukaryotic phytoplankton, bacterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service