All Photos(1)

Z803898

HPLC Autosampler Standards

certified reference material

grade

certified reference material

manufacturer/tradename

Jaytee Biosciences Ltd (CRM-1004)

Featured Industry

Food and Beverages
Petroleum
Pharmaceutical
Plastics

format

neat

General description

CRM for HPLC.

These HPLC autosampler standards are used to qualify HPLC autosamplers for reproducability, linearity and carryover.

Developed to be used in any laboratory regardless of the regulator and are as relevant to a pharmaceutical company as to a contract lab working to ISO 17025.

All products come supplied in flame-sealed ampoules to increase the shelf life and prevent contamination, with full certification.

Traceable instrument qualification adds confidence to all data generated. In addition, your audit trail is backed up by the independent verification by UKAS to ISO 17034.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK Germany

WGK 3

Flash Point F

91.4 °F

Flash Point C

33 °C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service