All Photos(1)

Z804126

UV Low Wavelength Standards

certified reference material

NACRES:
NA.21

grade

certified reference material

manufacturer/tradename

Jaytee Biosciences Ltd (CRM-1002)

Featured Industry

Food and Beverages
Petroleum
Pharmaceutical
Plastics

format

neat

General description

CRM for HPLC.

These UV low wavelength standards are designed to qualify UV/DAD detectors in the low UV region.

Developed to be used in any laboratory regardless of the regulator and are as relevant to a pharmaceutical company as to a contract lab working to ISO 17025.

All products come supplied in flame-sealed ampoules to increase the shelf life and prevent contamination, with full certification.

Traceable instrument qualification adds confidence to all data generated. In addition, your audit trail is backed up by the independent verification by UKAS to ISO 17034.

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Ox. Liq. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service