All Photos(1)

Z804223

UV Multi Wavelength Standards

certified reference material

NACRES:
NA.21

grade

certified reference material

manufacturer/tradename

Jaytee Biosciences Ltd (CRM-1001)

General description

CRM for HPLC.

These UV multi wavelength standards are designed to qualify UV/DAD detector wavelength up to 14 wavelengths.

Developed to be used in any laboratory regardless of the regulator and are as relevant to a pharmaceutical company as to a contract lab working to ISO 17025.

All products come supplied in flame-sealed ampoules to increase the shelf life and prevent contamination, with full certification.

Traceable instrument qualification adds confidence to all data generated. In addition, your audit trail is backed up by the independent verification by UKAS to ISO 17034.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service