Skip to Content
Merck

Skip To

76696

BioStab Alkaline Phosphatase Conjugate Stabilizer

Synonym(s):

Alkaline Phosphatase Conjugate Stabilizer BioStab

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12352202
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

technique(s)

HPLC: suitable

impurities

PHOSPHATASES, none detected, PROTEASES, none detected

suitability

corresponds for identity (HPLC)

storage temp.

2-8°C

General description

BioStab Alkaline Phosphatase Conjugate Stabilizer dramatically extends the life of AP conjugates, allowing more consistent results over extended periods of time compared to daily prepared dilutions. The mode of action of BioStab AP Stabilizer comprises both stabilization of the antibody and stabilization of the active center of AP.

Application

BioStab Alkaline Phosphatase Conjugate Stabilizer can be used in ELISA, Western Blotting, immunohistochemistry and any other application which requires AP conjugates. The stabilizer is free of any protein to minimize potential nonspecific interaction. BioStab Alkaline Phosphatase Conjugate Stabilizer can be used to dilute the conjugate to ready-to-use concentrations (below 20 ng/ml).


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters