Merck
All Photos(2)

CLS356239

Corning® Matrigel® Growth Factor Reduced (GFR) Basement Membrane Matrix

Phenol Red-Free, LDEV-Free, pack of 100 mL (10x10 mL)

feature

Phenol Red-Free, LDEV-Free

packaging

pack of 100 mL (10x10 mL)

manufacturer/tradename

Corning 356239

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

1 of 4

This Item
CLS356230CLS356230CLS356231
feature

Phenol Red-Free, LDEV-Free

feature

LDEV-Free

feature

LDEV-Free

feature

Phenol Red-Free, LDEV-Free

packaging

pack of 100 mL (10x10 mL)

packaging

pack of 5 mL

packaging

pack of 5 mL

packaging

pack of 10 mL

manufacturer/tradename

Corning 356239

manufacturer/tradename

Corning 356230

manufacturer/tradename

Corning 356230

manufacturer/tradename

Corning 356231

General description

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Quality

Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
Extensive PCR testing is performed to screen for a number of pathogens
Tested and found negative for bacteria, fungi, and mycoplasma
Protein concentrations determined by Lowry method
Endotoxin units measured by Limulus Amoebocyte Lysate assay
Matrigel Matrix gel stability is tested for a period of 14 days at 37°C
Biological activity is determined for each lot using a neurite outgrowth assay.
Source
Engelbreth-Holm-Swarm Mouse Tumor

Preparation and Storage
Store at -20°C. Avoid multiple freeze-thaws. Do not store in frost-free freezer. KEEP FROZEN.

Application

Cell Growth and Differentiation
Metabolism/Toxicology Studies
Invasion Assays
in vitro and in vivo Angiogenesis Assays
in vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice
Click here for Corning Matrigel Lot Selection and Reservation.

For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.

Legal Information

Corning is a registered trademark of Corning, Inc.
Matrigel is a registered trademark of Corning, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service